Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

OR61067-1G OR61067-5G
76594-030EA 29 USD InStock
76594-030 76594-028
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride


Formula: C₈H₁₅NO₂
MW: 193.67 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD08274538
CAS Number: 61367-07-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty