Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1H-pyrazole 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 3-Pyrazolamine1H-Pyrazol-3-amine3-Aminopyrazole3-AP

OR6125-100G OR6125-25G OR6125-5G
76621-770EA 36 USD InStock
76621-770 76621-772 76621-768
3-Amino-1H-pyrazole 97%
3-Amino-1H-pyrazole


Formula: C₃H₅N₃
MW: 83.09 g/mol
Boiling Pt: 218 °C/122mmHg
Melting Pt: 33…39 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
UN: 3263
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty