Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-(Boc-amino)piperidine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Aminopiperidine, 3-BOC protectedtert-Butyl piperidin-3-ylcarbamate

OR6133-5G OR6133-25G OR6133-1G
76617-136EA 101.97 USD InStock
76617-136 76617-144 76617-134
(±)-3-(Boc-amino)piperidine 97%
(±)-3-(Boc-amino)piperidine


Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 102…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839941
CAS Number: 172603-05-3
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty