Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salicylic acid 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: o-Hydroxybenzoic acid2-Hydroxybenzoic acid

OR6245-500G OR6245-250G OR6245-100G
76594-590EA 38 USD InStock
76594-590 76594-586 76594-588
Salicylic acid 99%
Salicylic acid


Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211 °C/20mmHg
Melting Pt: 158…161 °C
Density: 1.44 g/cm³ (25 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty