Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-3-hydroxypyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Bromo-3-pyridinol

OR6410-5G OR6410-100G OR6410-10G
76594-732EA 18.54 USD InStock
76594-732 76594-736 76594-734
2-Bromo-3-hydroxypyridine 98%
2-Bromo-3-hydroxypyridine


Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty