Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-Methylpyridin-4-amine2-Methyl-4-pyridylamine4-Amino-2-picoline

OR6421-5G OR6421-25G
76594-754EA 71 USD InStock
76594-754 76594-752
4-Amino-2-methylpyridine 97%
4-Amino-2-methylpyridine


Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00186509
CAS Number: 18437-58-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty