Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (4-Chlorophenyl)metaboric acidDihydroxy(4-chlorophenyl)borane(4-Chlorophenyl)boronic acid4-Chlorobenzeneboronic acid

OR6643-100G OR6643-25G OR6643-500G
76595-010EA 250 USD InStock
76595-010 76595-006 76595-008
4-Chlorophenylboronic acid 98%
4-Chlorophenylboronic acid


Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 284…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty