Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: p-Iodobenzoyl chloride

OR6727-100G OR6727-25G OR6727-250G
76621-840EA 35 USD InStock
76621-840 76621-836 76621-838
4-Iodobenzoyl chloride 97%
4-Iodobenzoyl chloride


Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 120…121 °C/1mmHg
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00001058
CAS Number: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty