Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver bromide

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Argentous bromide

OR7534-25G OR7534-100G OR7534-5G
76617-328EA 13 USD InStock
76617-328 76617-324 76617-326
Silver bromide
Silver bromide


Formula: AgBr
MW: 187.77 g/mol
Melting Pt: 432 °C
Storage Temperature: Ambient
MDL Number: MFCD00003398
CAS Number: 7785-23-1
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty