Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Boc-4-(4-bromophenyl)piperazine4-(4-Bromophenyl)piperazine-1-carboxylic acid tert-Butyl ester1-tert-Butoxycarbonyl-4-(4-bromophenyl)piperazine

OR8542-1G OR8542-5G
76588-832EA 101 USD InStock
76588-832 76588-830
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(4-Bromophenyl)piperazine-1-carboxylate


Formula: C₁₅H₂₁BrN₂O₂
MW: 341.25 g/mol
Melting Pt: 146…147 °C
Storage Temperature: Ambient
MDL Number: MFCD07371649
CAS Number: 352437-09-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty