Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Picolyl chloride hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-(Chloromethyl)pyridine hydrochloride

OR8686-5G OR8686-25G OR8686-100G
76621-924EA 13.39 USD InStock
76621-924 76621-920 76621-922
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride


Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 119…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00012811
CAS Number: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty