Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-4-iodotoulene 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-(Bromomethyl)-4-iodobenzene4-Iodobenzyl bromideα-Bromo-p-iodotoulene

OR8750-5G OR8750-25G OR8750-100G
76621-930EA 16 USD InStock
76621-930 76621-928 76621-926
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene


Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 140 °C/15mmHg
Melting Pt: 78…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty