Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrachloro-p-benzoquinone 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Tetrachloroquinonep-Chloranil2,3,5,6-Tetrachlorobenzo-1,4-quinone2,3,5,6-Tetrachloro-1,4-benzoquinoneTetrachloro-1,4-benzoquinone4-Chloranil

OR8777-25G OR8777-500G OR8777-100G
76617-428EA 28 USD InStock
76617-428 76617-430 76617-432
Tetrachloro-p-benzoquinone 98%
Tetrachloro-p-benzoquinone


Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Melting Pt: 287…290 °C
Density: 1.97 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty