Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dichloropyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR8832-10G OR8832-5G
76589-128EA 14 USD InStock
76589-128 76589-126
3,4-Dichloropyridine
3,4-Dichloropyridine


Formula: C₅H₃Cl₂N
Melting Pt: 22…23 °C
Storage Temperature: Ambient
MDL Number: MFCD01861989
CAS Number: 55934-00-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty