Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-bromo-4-hydroxybenzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 3-bromo-4-hydroxybenzoate

OR908176-25G
76589-476EA 45 USD InStock
76589-476
Methyl-3-bromo-4-hydroxybenzoate 98%
Methyl 3-bromo-4-hydroxybenzoate


Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203850
CAS Number: 29415-97-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty