Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylethylene

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1,1,2-Triphenylethylene1,1,2-Triphenylethene

OR9099-250G OR9099-50G OR9099-10G
76617-460EA 227 USD InStock
76617-460 76617-458 76617-462
Triphenylethylene
Triphenylethylene


Formula: (C₆H₅)₂CCHC₆H₅
MW: 256.35 g/mol
Boiling Pt: 219…221 °C/14mmHg
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004765
CAS Number: 58-72-0
EINECS: 200-395-1
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty