Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl-3-oxocyclobutanecarboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: tert-Butyl 3-oxocyclobutanecarboxylate

OR921343-25G OR921343-10G OR921343-5G
76589-766EA 75 USD InStock
76589-766 76589-762 76589-764
tert-Butyl-3-oxocyclobutanecarboxylate 95%
tert-Butyl-3-oxocyclobutanecarboxylate


Formula: C₉H₁₄O₃
MW: 170.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827541
CAS Number: 145549-76-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty