Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N1,N2-Dimethylcyclohexane-1,2-diamine 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR928538-5G
76589-900EA 42 USD InStock
76589-900
N1,N2-Dimethylcyclohexane-1,2-diamine 96%
N1,N2-Dimethylcyclohexane-1,2-diamine


Formula: C₈H₁₈N₂
MW: 142.2419 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664382
CAS Number: 61798-24-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty