Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Prop-2-ynoxybenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR945348-5G OR945348-250MG OR945348-1G
76590-212EA 26 USD InStock
76590-212 76590-210 76590-214
3-Prop-2-ynoxybenzaldehyde 98%
3-Prop-2-ynoxybenzaldehyde


Formula: C₁₀H₈O₂
MW: 160.169 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05625818
CAS Number: 5651-87-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty