Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-chloro-1,2,4-thiadiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR9538-5G OR9538-10G OR9538-1G
76590-448EA 240 USD InStock
76590-448 76590-446 76590-444
3-Bromo-5-chloro-1,2,4-thiadiazole
3-Bromo-5-chloro-1,2,4-thiadiazole


Formula: C₂BrClN₂S
MW: 199.45 g/mol
Boiling Pt: 192…194 °C
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00799287
CAS Number: 37159-60-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty