Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dihydroxybenzoic acid, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,5-DHBA2,5-Dihydroxybenzoic AcidGentisic acidHydroquinonecarboxylic acidDHB

OR9586-25G OR9586-100G
76590-552EA 43 USD InStock
76590-552 76590-550
2,5-Dihydroxybenzoic acid, Technical Grade
2,5-Dihydroxybenzoic acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9
EINECS: 207-718-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty