Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethyl-3-iodo-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR959875-5G OR959875-1G
76590-604EA 41 USD InStock
76590-604 76590-606
1-Ethyl-3-iodo-1H-pyrazole
1-Ethyl-3-iodo-1H-pyrazole


Formula: C₅H₇IN₂
Storage Temperature: Ambient
MDL Number: MFCD12756564
CAS Number: 1202781-34-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty