Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenylbenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Biphenylcarboxaldehydep-Biphenylylaldehyde[1,1'-Biphenyl]-4-carbaldehyde4-Biphenylcarbaldehyde4-BiphenylylaldehydeBiphenyl-4-carboxaldehydeBiphenyl-4-carbaldehyde

OR9875-25G OR9875-500G OR9875-100G
76590-858EA 38 USD InStock
76590-858 76590-856 76590-860
4-Phenylbenzaldehyde 98%
4-Phenylbenzaldehyde


Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 184 °C/11mmHg
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00006947
CAS Number: 3218-36-8
EINECS: 221-742-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty