Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-nitrobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC1197-5G PC1197-100G PC1197-25G
76625-702EA 14 USD InStock
76625-702 76625-700 76625-698
4-Fluoro-3-nitrobenzonitrile
4-Fluoro-3-nitrobenzonitrile


Formula: C₇H₃FN₂O₂
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01632197
CAS Number: 1009-35-4
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty