Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-1,2-difluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Bromo-3,4-difluorobenzene

PC1383-50G PC1383-500G PC1383-250G
76612-810EA 13.39 USD InStock
76612-810 76612-808 76612-806
4-Bromo-1,2-difluorobenzene 98%
4-Bromo-1,2-difluorobenzene


Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 150…151 °C
Melting Pt: –4 °C
Density: 1.707 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00000304
CAS Number: 348-61-8
EINECS: 206-481-5
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty