Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-fluoroaniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1819A-25G PC1819A-5G
76612-902EA 13.39 USD InStock
76612-902 76612-900
2-Chloro-5-fluoroaniline 97%
2-Chloro-5-fluoroaniline


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00084852
CAS Number: 452-83-5
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty