Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-bromo-6-fluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 6-Amino-3-bromo-2-fluorobenzonitrile

PC201321-25G PC201321-1G PC201321-5G
76625-758EA 38 USD InStock
76625-758 76625-756 76625-754
2-Amino-5-bromo-6-fluorobenzonitrile 95%
2-Amino-5-bromo-6-fluorobenzonitrile


Formula: C₇H₄BrFN₂
MW: 215.0225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657979
CAS Number: 845866-92-4
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty