Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) fluoride, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC2100-5G PC2100-25G PC2100-100G
77413-530EA 41 USD InStock
77413-530 76617-834 76617-832
Copper(II) fluoride, anhydrous 99%
Copper (II) fluoride


Formula: CuF₂
MW: 101.54 g/mol
Melting Pt: 950 °C (dec.)
Density: 4.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS Number: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty