Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Difluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC2620-500G PC2620-1KG PC2620-2.5KG
76613-028EA 265 USD InStock
76613-028 76613-030 76613-032
1,2-Difluorobenzene 99%
1,2-Difluorobenzene


Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 92 °C
Melting Pt: –34 °C
Density: 1.158 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000284
CAS Number: 367-11-3
EINECS: 206-680-7
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty