Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluorobenzenesulfonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,4-Difluorobenzene-1-sulfonyl chloride2,4-Difluorobenzene-1-sulphonyl chloride

PC2644-25G PC2644-5G PC2644-100G
76622-488EA 69 USD InStock
76622-488 76622-492 76622-490
2,4-Difluorobenzenesulfonyl chloride 98%
2,4-Difluorobenzenesulphonyl chloride


Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 101…102 °C/1mmHg
Density: 1.581 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty