Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Difluorobenzophenone 98+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Bis(4-Fluorophenyl)methanoneBis(4-fluorophenyl) ketone

PC2696-250G PC2696-1KG
76617-896EA 95 USD InStock
76617-896 76617-898
4,4'-Difluorobenzophenone 98+%
4,4'-Difluorobenzophenone
Intermediate for the manufacture of polyetheretherketone(PEEK) high performance polymer.


Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 170…172 °C/10mmHg
Melting Pt: 102…110 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty