Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodo-2-(trifluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC2697-5G PC2697-25G
76625-812EA 18 USD InStock
76625-812 76625-810
4-Iodo-2-(trifluoromethyl)benzonitrile
4-Iodo-2-(trifluoromethyl)benzonitrile


Formula: C₈H₃F₃IN
MW: 297.02 g/mol
Melting Pt: 50…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08458091
CAS Number: 101066-87-9
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty