Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Difluoro-4-nitrobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,4-Difluoro-1-nitrobenzene2,4-Difluoronitrobenzene

PC2850-500G PC2850-100G PC2850-1KG
76613-072EA 13 USD InStock
76613-072 76613-074 76613-076
1,3-Difluoro-4-nitrobenzene 99%
1,3-Difluoro-4-nitrobenzene


Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 203…204 °C
Melting Pt: 10 °C
Density: 1.451 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007050
CAS Number: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty