Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Difluorophenyl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Isocyanic acid 3,4-Difluorophenyl ester

PC2875K-5G PC2875K-25G PC2875K-1G
76613-080EA 114 USD InStock
76613-080 76613-082 76613-078
3,4-Difluorophenyl isocyanate 97%
3,4-Difluorophenyl isocyanate
  • May form small amount of sediment.


Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 164 °C
Density: 1.326 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065711
CAS Number: 42601-04-7
UN: 2206
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty