Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Difluorophenyl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1,4-Difluoro-2-isocyanatobenzene

PC2875I-1G PC2875I-5G
76617-932EA 45 USD InStock
76617-932 76617-930
2,5-Difluorophenyl isocyanate 97%
2,5-Difluorophenyl isocyanate
  • May form small amount of sediment.


Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 78 °C/33mmHg
Density: 1.323 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013843
CAS Number: 39718-32-6
UN: 3080
ADR: 6.1+3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty