Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dichloro-6-(trifluoromethyl)benzoic acid 98%

Supplier: APOLLO SCIENTIFIC

PC302655-1G PC302655-5G PC302655-10G
76598-320EA 24 USD InStock
76598-320 76598-322 76598-318
2,3-Dichloro-6-(trifluoromethyl)benzoic acid 98%
2,3-Dichloro-6-(trifluoromethyl)benzoic acid


Formula: C₈H₃Cl₂F₃O₂
MW: 259.01 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD06660270
CAS Number: 25922-43-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty