Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,6-Difluoro-m-toluic acid 97%

Supplier: APOLLO SCIENTIFIC
Synonyms: 2,4-Difluoro-5-methylbenzoic acid

PC303306-1G PC303306-5G
76599-614EA 23 USD InStock
76599-614 76599-616
4,6-Difluoro-m-toluic acid 97%
4,6-Difluoro-m-toluic acid


Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Melting Pt: 159…166 °C
Storage Temperature: Ambient
MDL Number: MFCD09835285
CAS Number: 367954-99-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty