Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxy-2,4-difluorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC302722-25G PC302722-5G
76598-520EA 256 USD InStock
76598-520 76598-518
3-Ethoxy-2,4-difluorobenzoic acid
3-Ethoxy-2,4-difluorobenzoic acid


Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09258723
CAS Number: 1017779-88-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty