Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-n-Butoxy-2,6-difluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Synonyms: 4-Butoxy-2,6-difluorobenzaldehyde

PC302736-1G PC302736-10G PC302736-5G
76598-554EA 118 USD InStock
76598-554 76598-550 76598-552
4-n-Butoxy-2,6-difluorobenzaldehyde 98%
4-n-Butoxy-2,6-difluorobenzaldehyde


Formula: C₁₁H₁₂F₂O₂
MW: 214.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201017
CAS Number: 1373920-93-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty