Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-methyl-DL-phenylalanine

Supplier: APOLLO SCIENTIFIC
Warning

PC302151-1G PC302151-10G PC302151-5G
76597-026EA 429 USD InStock
76597-026 76597-024 76597-022
4-Fluoro-3-methyl-DL-phenylalanine
4-Fluoro-3-methyl-DL-phenylalanine


Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116031
CAS Number: 1259956-32-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty