Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-4-ethoxy-5-fluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC

PC302945-10G PC302945-5G PC302945-1G
76622-992EA 352 USD InStock
76622-992 76622-994 76622-996
3-Chloro-4-ethoxy-5-fluorobenzyl bromide 98%
3-Chloro-4-ethoxy-5-fluorobenzyl bromide


Formula: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832342
CAS Number: 1017778-91-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty