Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%

Supplier: APOLLO SCIENTIFIC

PC303350-1G PC303350-5G
76613-632EA 196 USD InStock
76613-632 76613-634
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline 97%
2-Bromo-5-chloro-4-(trifluoromethoxy)aniline


Formula: C₇H₄BrClF₃NO
MW: 290.465 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28734569
CAS Number: 868406-81-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty