Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline 97%

Supplier: APOLLO SCIENTIFIC
Synonyms: 3-Fluoro-4-(3-chloro-2-fluorophenoxy)aniline

PC303106-5G PC303106-10G PC303106-1G
76613-478EA 63 USD InStock
76613-478 76613-476 76613-480
4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline 97%
4-(3-Chloro-2-fluorophenoxy)-3-fluoroaniline


Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty