Would you like to log in again?
ESD Stacking Bins
Supplier: Treston
1520-4ESD 1015-4ESD 3050-4ESD 1525-4ESD 1950-4ESD 3040-4ESD SBS4-2ESD 1930-4ESD 1940-4ESD SBS4-1015-4ESD
76015-034PK 86.45 USD InStock
76015-034 10496-567 10496-569 10682-749 10486-477 10682-747 10682-745 10496-571 76015-032 10682-743
ESD Stacking Bins
Boxes ESD-Safe Boxes
These high quality, durable stacking bins made from semiconductive polypropylene (PP) have been designed for daily use in stores, production facilities and warehouses.
  • Bins of the same width stack securely, even if they are of different depths
  • Straight sides ensure an efficient use of space
  • Withstand the effects of oils, most acids and solvents
  • Suitable for use in cleanroom environments
Bins have been designed to fit all standard metric shelves, paternoster and industrial workstations. The design, with a half-open front, allows excellent visibility and easy access to the bin's contents, making it easy to pick from.
Ordering information: Labels and dividers sold separately. Installation is required. Assembly videos available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty