Would you like to log in again?
PYREX® Stopcocks Keys, Corning
Supplier: Corning
PYREX®
7681-4 7681-6 7681-2 7681-8
29610-634CS 120.7 USD InStock
29610-634 29610-638 29610-640 29610-642
PYREX® Stopcocks Keys, Corning
Stopcock Keys
These are replacement PTFE stopcock plug assemblies with various diameter bores for PYREX® straight bore PTFE stopcocks.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty