Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Perfluorobenzene

PC4630-500G PC4630-250G PC4630-50G
76613-998EA 189 USD InStock
76613-998 76614-002 76614-000
Hexafluorobenzene 99%
Hexafluorobenzene


Formula: C₆F₆
MW: 186.06 g/mol
Boiling Pt: 80…82 °C
Melting Pt: 44351 °C
Density: 1.612 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000288
CAS Number: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty