Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3460-1KG PC3460-500G PC3460-100G
76613-768EA 120 USD InStock
76613-768 76613-766 76613-770
3-Fluorobenzaldehyde 98%
3-Fluorobenzaldehyde


Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 56…58 °C/10mmHg
Density: 1.174 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty