Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-p-anisaldehyde

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Fluoro-4-anisaldehyde3-Fluoro-4-methoxybenzaldehyde

PC5655-5G PC5655-25G
76602-506EA 45 USD InStock
76602-506 76602-508
3-Fluoro-p-anisaldehyde
3-Fluoro-p-anisaldehyde


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 129…132 °C/11mmHg
Melting Pt: 34…35 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003349
CAS Number: 351-54-2
EINECS: 206-514-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty