Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-3-fluorotoulene 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-(Chloromethyl)-3-fluorobenzene3-Fluorobenzyl chlorideα-Chloro-m-fluorotoulene

PC3710-100G PC3710-1KG
76618-218EA 105 USD InStock
76618-218 76618-216
α-Chloro-3-fluorotoulene 97%
α-Chloro-3-fluorotoulene


Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 84 °C/29mmHg
Density: 1.213 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00000904
CAS Number: 456-42-8
EINECS: 207-264-8
UN: 2920
ADR: 8+3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty