Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-3-tolualdehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Cinacalcet Impurity 233-(Trifluoromethyl)benzaldehyde

PC7500-100G PC7500-500G PC7500-25G
76618-904EA 24 USD InStock
76618-904 76618-908 76618-906
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde


Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83…86 °C/30mmHg
Density: 1.304 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
UN: 3082
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty